Product Details

SNP ID
rs1059331
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17165055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTCTCTCATCACACACATACA[C/T]ACTCAGTCTTGCTGAGGGAGAATTC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CECR5 PubMed Links

Gene Details

Gene
CECR5
Gene Name
cat eye syndrome chromosome region, candidate 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017829.5 Intron NP_060299.4
NM_033070.2 Intron NP_149061.1
XM_005261236.3 Intron XP_005261293.1
XM_011546126.2 Intron XP_011544428.1
XM_011546127.1 Intron XP_011544429.1
XM_017028781.1 Intron XP_016884270.1
XM_017028782.1 Intron XP_016884271.1
XM_017028783.1 Intron XP_016884272.1
XM_017028784.1 Intron XP_016884273.1
Gene
CECR5-AS1
Gene Name
CECR5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details