Product Details

SNP ID
rs928831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17043313 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAAAGGCACTCACGTTGATCTAG[C/T]ATCACAAGTGGAAAACAGTATTACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CECR7 PubMed Links

Gene Details

Gene
CECR7
Gene Name
cat eye syndrome chromosome region, candidate 7 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details