Product Details

SNP ID
rs1046929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101628397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAGAGTGAAGGGTTGCAAAAGAA[A/C]TGTAAGGGACTATGGGCAGCTGGGG
Phenotype
MIM: 300364
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARMCX3 PubMed Links

Gene Details

Gene
ARMCX3
Gene Name
armadillo repeat containing, X-linked 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016607.3 4080 Intron NP_057691.1
NM_177947.2 4080 Intron NP_808816.1
NM_177948.2 4080 Intron NP_808817.1
XM_005262141.2 4080 UTR 3 XP_005262198.1

View Full Product Details