Product Details

SNP ID
rs3852699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67227622 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATTGAATTGTTCCAGGGCGAGCA[A/G]GAGGCGATTGATCCTAGCTGCGGGT
Phenotype
MIM: 606881
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FHOD1 PubMed Links

Gene Details

Gene
FHOD1
Gene Name
formin homology 2 domain containing 1
There are no transcripts associated with this gene.

Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011523526.2 Intron XP_011521828.1
Gene
LRRC29
Gene Name
leucine rich repeat containing 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004055.1 Intron NP_001004055.1
NM_012163.2 Intron NP_036295.1
XM_017023126.1 Intron XP_016878615.1
XM_017023127.1 Intron XP_016878616.1
XM_017023128.1 Intron XP_016878617.1
XM_017023129.1 Intron XP_016878618.1
XM_017023130.1 Intron XP_016878619.1
XM_017023131.1 Intron XP_016878620.1
XM_017023132.1 Intron XP_016878621.1
XM_017023133.1 Intron XP_016878622.1
XM_017023134.1 Intron XP_016878623.1
Gene
TMEM208
Gene Name
transmembrane protein 208
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318217.1 Intron NP_001305146.1
NM_014187.3 Intron NP_054906.2

View Full Product Details