Product Details

SNP ID
rs868213
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:67186554 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAGGAATGGGTCAGTGCCTCAGCA[A/G]ACCTCCTCAAGAATGTGCCCAGTTC
Phenotype
MIM: 600659 MIM: 614117
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
E2F4 PubMed Links

Gene Details

Gene
E2F4
Gene Name
E2F transcription factor 4
There are no transcripts associated with this gene.

Gene
EXOC3L1
Gene Name
exocyst complex component 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178516.3 Intron NP_848611.2
Gene
KIAA0895L
Gene Name
KIAA0895 like
There are no transcripts associated with this gene.

View Full Product Details