Product Details

SNP ID
rs1544888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58059544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGGCGGGGCCCAACGCCCAGC[G/T]GGACAGGAGCTGCTCCGACGGCCCC
Phenotype
MIM: 604077
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF135 PubMed Links

Gene Details

Gene
ZNF135
Gene Name
zinc finger protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164527.1 134 Intron NP_001157999.1
NM_001164529.1 134 Intron NP_001158001.1
NM_001164530.1 134 Intron NP_001158002.1
NM_001289401.1 134 Intron NP_001276330.1
NM_001289402.1 134 Intron NP_001276331.1
NM_003436.3 134 Intron NP_003427.3
NM_007134.1 134 Intron NP_009065.1
XM_005259211.1 134 Intron XP_005259268.1
XM_006723362.3 134 Intron XP_006723425.1
XM_006723363.3 134 UTR 5 XP_006723426.1
XM_017027240.1 134 UTR 5 XP_016882729.1
XM_017027241.1 134 Intron XP_016882730.1
XM_017027242.1 134 Intron XP_016882731.1
Gene
ZSCAN1
Gene Name
zinc finger and SCAN domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182572.3 134 Intron NP_872378.3
XM_006723149.2 134 Intron XP_006723212.1
XM_017026609.1 134 Intron XP_016882098.1
XM_017026610.1 134 Intron XP_016882099.1

View Full Product Details