Product Details
- SNP ID
-
rs1414929
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:93569059 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCCACGCAGATGCCTCTAAAAGCA[A/T]GGATCAGGAGGGGGCTGGGCTTCTG
- Phenotype
-
MIM: 609044
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FFAR4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76035592] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FFAR4
- Gene Name
- free fatty acid receptor 4
View Full Product Details