Product Details
- SNP ID
-
rs1298034
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:76669233 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAACACTGGGAGTTACATTTCAAC[A/C]TGAAATTTTGAGGATGCAAATATCC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR325HG
PubMed Links
Gene Details
- Gene
- MIR325HG
- Gene Name
- MIR325 host gene
There are no transcripts associated with this gene.
View Full Product Details