Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_033034.2 | Intron | NP_149023.2 | |||
NM_033092.2 | Intron | NP_149083.2 | |||
NM_033093.2 | Intron | NP_149084.2 | |||
XM_005253183.2 | Intron | XP_005253240.1 | |||
XM_005253184.2 | Intron | XP_005253241.1 | |||
XM_006718358.2 | Intron | XP_006718421.1 | |||
XM_011520426.2 | Intron | XP_011518728.1 | |||
XM_011520427.1 | Intron | XP_011518729.1 | |||
XM_017018460.1 | Intron | XP_016873949.1 | |||
XM_017018461.1 | Intron | XP_016873950.1 | |||
XM_017018462.1 | Intron | XP_016873951.1 | |||
XM_017018463.1 | Intron | XP_016873952.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001003818.2 | Intron | NP_001003818.1 | |||
NM_001198644.1 | Intron | NP_001185573.1 | |||
NM_001198645.1 | Intron | NP_001185574.1 | |||
NM_058166.4 | Intron | NP_477514.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001003819.3 | Intron | NP_001003819.1 |