Product Details

SNP ID
rs11758502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138904654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATTAAATGGGAAACTCTTTCT[A/G]AAGATTTTTATAGAATAGATGCTTT
Phenotype
MIM: 614825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECT2L PubMed Links

Gene Details

Gene
ECT2L
Gene Name
epithelial cell transforming 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077706.2 3107 Intron NP_001071174.1
NM_001195037.2 3107 Intron NP_001181966.1
XM_006715472.3 3107 Intron XP_006715535.1
XM_011535795.2 3107 Intron XP_011534097.1
XM_011535797.2 3107 Intron XP_011534099.1
XM_017010828.1 3107 Intron XP_016866317.1
XM_017010829.1 3107 Intron XP_016866318.1
XM_017010830.1 3107 Intron XP_016866319.1
Gene
REPS1
Gene Name
RALBP1 associated Eps domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128617.2 3107 UTR 3 NP_001122089.1
NM_001286611.1 3107 UTR 3 NP_001273540.1
NM_001286612.1 3107 UTR 3 NP_001273541.1
NM_031922.4 3107 UTR 3 NP_114128.3
XM_005267177.3 3107 Intron XP_005267234.1
XM_005267178.4 3107 UTR 3 XP_005267235.1
XM_005267179.3 3107 Intron XP_005267236.1
XM_011536202.2 3107 Intron XP_011534504.1
XM_017011387.1 3107 Intron XP_016866876.1
XM_017011388.1 3107 UTR 3 XP_016866877.1
XM_017011389.1 3107 Intron XP_016866878.1

View Full Product Details