Product Details
- SNP ID
-
rs8059717
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56431513 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCAGGAAAATAGCCCTGTGACTGG[C/T]ACCATACTAATCAAAAGTTCATTTC
- Phenotype
-
MIM: 603243
MIM: 604978
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMFR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78002423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMFR
- Gene Name
- autocrine motility factor receptor
There are no transcripts associated with this gene.
- Gene
- NUDT21
- Gene Name
- nudix hydrolase 21
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007006.2 |
2030 |
UTR 3 |
|
|
NP_008937.1 |
View Full Product Details