Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_182496.2 | 1626 | Silent Mutation | TTC,TTT | F533F | NP_872302.2 |
XM_006719229.2 | 1626 | Silent Mutation | TTC,TTT | F369F | XP_006719292.1 |
XM_011537883.2 | 1626 | Silent Mutation | TTC,TTT | F533F | XP_011536185.1 |
XM_011537884.2 | 1626 | Silent Mutation | TTC,TTT | F493F | XP_011536186.1 |
XM_011537886.2 | 1626 | Silent Mutation | TTC,TTT | F493F | XP_011536188.1 |
XM_011537887.2 | 1626 | Silent Mutation | TTC,TTT | F493F | XP_011536189.1 |
XM_011537888.2 | 1626 | Silent Mutation | TTC,TTT | F316F | XP_011536190.1 |
XM_011537889.1 | 1626 | Intron | XP_011536191.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003095.2 | 1626 | Intron | NP_003086.1 |