Product Details

SNP ID
rs12917724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1257295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTGGAGAGTCCCTTAGCACCTC[C/T]GTGCCTCGGTTTCCCCTTGCCTGAA
Phenotype
MIM: 609272
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS29P PubMed Links

Gene Details

Gene
PRSS29P
Gene Name
protease, serine 29, pseudogene
There are no transcripts associated with this gene.

Gene
TPSD1
Gene Name
tryptase delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012217.2 Intron NP_036349.1

View Full Product Details