Product Details

SNP ID
rs12462359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52032168 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTACATAATGGTACCTAGTAACT[G/T]GGATAAACACATCCAAATATACTGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF432 PubMed Links

Gene Details

Gene
ZNF432
Gene Name
zinc finger protein 432
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322284.1 4133 UTR 3 NP_001309213.1
NM_001322285.1 4133 UTR 3 NP_001309214.1
NM_014650.3 4133 UTR 3 NP_055465.1
Gene
ZNF614
Gene Name
zinc finger protein 614
There are no transcripts associated with this gene.

View Full Product Details