Product Details

SNP ID
rs7750700
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:49467981 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGATTTGATTAAGGAGTAGTAGGA[A/G]AGAAGTATGAAAAGGTAGGCAGAGT
Phenotype
MIM: 611506 MIM: 609058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPQ PubMed Links

Gene Details

Gene
CENPQ
Gene Name
centromere protein Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018132.3 Intron NP_060602.2
XM_005249205.1 Intron XP_005249262.1
XM_011514723.2 Intron XP_011513025.2
Gene
MUT
Gene Name
methylmalonyl-CoA mutase
There are no transcripts associated with this gene.

View Full Product Details