Product Details

SNP ID
rs2569448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51013190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGGAAGGGAGCTGTGTGCTCAGAC[A/C]TTGTACATCAACCTCCTGTGGGCTC
Phenotype
MIM: 602673 MIM: 604434 MIM: 605504
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLK10 PubMed Links

Gene Details

Gene
KLK10
Gene Name
kallikrein related peptidase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077500.1 2561 UTR 3 NP_001070968.1
NM_002776.4 2561 UTR 3 NP_002767.2
NM_145888.2 2561 UTR 3 NP_665895.1
XM_005259061.3 2561 Intron XP_005259118.1
XM_005259062.3 2561 Intron XP_005259119.1
XM_006723287.3 2561 Intron XP_006723350.1
XM_006723289.3 2561 Intron XP_006723352.1
XM_017026993.1 2561 Intron XP_016882482.1
Gene
KLK11
Gene Name
kallikrein related peptidase 11
There are no transcripts associated with this gene.

Gene
KLK9
Gene Name
kallikrein related peptidase 9
There are no transcripts associated with this gene.

View Full Product Details