Product Details

SNP ID
rs12208531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:115942361 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGATAACATTGTATTTTGGAAT[A/G]GATTATTTGAATTTGTTTTGCTACT
Phenotype
MIM: 606573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRK PubMed Links

Gene Details

Gene
FRK
Gene Name
fyn related Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002031.2 2208 UTR 3 NP_002022.1
XM_005266880.4 2208 UTR 3 XP_005266937.1
XM_005266881.2 2208 UTR 3 XP_005266938.1
XM_005266882.4 2208 UTR 3 XP_005266939.1
XM_011535653.2 2208 UTR 3 XP_011533955.1
XM_011535654.2 2208 UTR 3 XP_011533956.1
XM_011535655.2 2208 UTR 3 XP_011533957.1
XM_011535656.2 2208 UTR 3 XP_011533958.1
XM_017010645.1 2208 UTR 3 XP_016866134.1

View Full Product Details