Product Details

SNP ID
rs16979231
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:44538778 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCTAAGGGTTCCATCTTTGAAAC[C/T]CATTAACCTTCTTTTCTGCCCCCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM20 PubMed Links

Gene Details

Gene
CEACAM20
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
There are no transcripts associated with this gene.

Gene
CEACAM22P
Gene Name
carcinoembryonic antigen related cell adhesion molecule 22, pseudogene
There are no transcripts associated with this gene.

View Full Product Details