Product Details

SNP ID
rs2894432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41209514 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATTTTTCCTGCCTCTGTGATCT[C/G]GCTTCTTTTTCCTAGCCATCTTTCT
Phenotype
MIM: 609715 MIM: 609716
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TREML2 PubMed Links

Gene Details

Gene
TREML2
Gene Name
triggering receptor expressed on myeloid cells like 2
There are no transcripts associated with this gene.

Gene
TREML3P
Gene Name
triggering receptor expressed on myeloid cells like 3, pseudogene
There are no transcripts associated with this gene.

View Full Product Details