Product Details
- SNP ID
-
rs11154302
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:125276681 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TCAGAGGGGCTAGGCTGGCTCATGA[G/T]GTAAACAAGGATGCAGGTGTCTCTC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HDDC2
PubMed Links
Gene Details
- Gene
- HDDC2
- Gene Name
- HD domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016063.2 |
|
Intron |
|
|
NP_057147.2 |
View Full Product Details