Product Details

SNP ID
rs35277048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26764943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTGTCTGAGACTGTAAGTCAAG[A/G]AAAGTCTACGGAAAGGCCAGATCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35F6 PubMed Links

Gene Details

Gene
SLC35F6
Gene Name
solute carrier family 35 member F6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017877.3 Intron NP_060347.2

View Full Product Details