Product Details

SNP ID
rs10865439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26789967 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCTTCCCTGGCTTCTTATTACA[G/T]TAGGATTAAATTCCATCTCCTCATG
Phenotype
MIM: 117139
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CENPA PubMed Links

Gene Details

Gene
CENPA
Gene Name
centromere protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042426.1 Intron NP_001035891.1
NM_001809.3 Intron NP_001800.1
Gene
SLC35F6
Gene Name
solute carrier family 35 member F6
There are no transcripts associated with this gene.

View Full Product Details