Product Details

SNP ID
rs4150847
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:85182766 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTCTACATTTTATTTGCCATTTTGG[T/C]GTCAGTATGAGCTTTTTTAAGCATG
Phenotype
MIM: 600967
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
E2F5 PubMed Links

Gene Details

Gene
E2F5
Gene Name
E2F transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083588.1 Intron NP_001077057.1
NM_001083589.1 Intron NP_001077058.1
NM_001951.3 Intron NP_001942.2
Gene
LOC102723322
Gene Name
uncharacterized LOC102723322
There are no transcripts associated with this gene.

View Full Product Details