Product Details
- SNP ID
-
rs11150847
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:80179331 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTCCACTGTGGTTATTCATATCATT[A/G]AATCCAGTGCACAGTTTAAAGGACC
- Phenotype
-
MIM: 607211
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CARD14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112891168] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CARD14
- Gene Name
- caspase recruitment domain family member 14
View Full Product Details