Product Details

SNP ID
rs7211994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80178715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGGCGCTGCTCCTGCACACAC[A/G]ACTCTCCAACGATCATTTTCCATGT
Phenotype
MIM: 607211
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARD14 PubMed Links

Gene Details

Gene
CARD14
Gene Name
caspase recruitment domain family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257970.1 Intron NP_001244899.1
NM_024110.4 Intron NP_077015.2
NM_052819.2 Intron NP_438170.1
XM_011525212.1 Intron XP_011523514.1
XM_011525213.1 Intron XP_011523515.1
XM_011525215.1 Intron XP_011523517.1
XM_011525216.1 Intron XP_011523518.1
XM_011525217.1 Intron XP_011523519.1
XM_011525218.2 Intron XP_011523520.1
XM_017025057.1 Intron XP_016880546.1
XM_017025058.1 Intron XP_016880547.1

View Full Product Details