Product Details
- SNP ID
-
rs11656113
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75591266 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGTGGTGAGTGGTGGCCTTCCTG[A/G]GTGGCTGAACCCATGGGCCACACTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYO15B
PubMed Links
Gene Details
- Gene
- MYO15B
- Gene Name
- myosin XVB
View Full Product Details