Product Details

SNP ID
rs2289503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100402277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTTGCAGAAGCCTTACATTAA[A/G]ATTTCATAAATGATTGTTTTGCTGT
Phenotype
MIM: 606081
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LNP1 PubMed Links

Gene Details

Gene
LNP1
Gene Name
leukemia NUP98 fusion partner 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085451.1 1085 UTR 5 NP_001078920.1
Gene
TOMM70
Gene Name
translocase of outer mitochondrial membrane 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014820.4 1085 Intron NP_055635.3

View Full Product Details