Product Details

SNP ID
rs306506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55948248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGAACTGTGATAAAATGTGTGTT[C/G]TAGTCCGCAGAGAGATAAATGGTGA
Phenotype
MIM: 609659
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NLRP8 PubMed Links

Gene Details

Gene
NLRP8
Gene Name
NLR family pyrin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317000.1 417 Missense Mutation CTA,GTA L116V NP_001303929.1
NM_176811.2 417 Missense Mutation CTA,GTA L116V NP_789781.2

View Full Product Details