Product Details
- SNP ID
-
rs7169976
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:73562702 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGGGCTAGACAGTTTATCCAGGCTA[C/T]AGTTCAGCCTCAAATTGCATTACAT
- Phenotype
-
MIM: 612820
MIM: 615176
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NPTN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28726349] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NPTN
- Gene Name
- neuroplastin
- Gene
- NPTN-IT1
- Gene Name
- NPTN intronic transcript 1
There are no transcripts associated with this gene.
- Gene
- REC114
- Gene Name
- REC114 meiotic recombination protein
There are no transcripts associated with this gene.
View Full Product Details