Product Details

SNP ID
rs2289608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:3248161 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTGAGCTTGGGCGACTGCCGGG[C/T]CCGCGAAACCCGACCCCTGCAGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927044 PubMed Links

Gene Details

Gene
LOC101927044
Gene Name
uncharacterized LOC101927044
There are no transcripts associated with this gene.

Gene
LOC104968399
Gene Name
uncharacterized LOC104968399
There are no transcripts associated with this gene.

Gene
MYL12A
Gene Name
myosin light chain 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303047.1 Intron NP_001289976.1
NM_001303048.1 Intron NP_001289977.1
NM_001303049.1 Intron NP_001289978.1
NM_006471.3 Intron NP_006462.1

View Full Product Details