Product Details
- SNP ID
-
rs4808209
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:19679350 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCGTCATTCATTCCTGGACAGGCAC[A/C]TGAGAGCTCATGCTGGACACAAACG
- Phenotype
-
MIM: 603983
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF101
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59327775] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF101
- Gene Name
- zinc finger protein 101
View Full Product Details