Product Details
- SNP ID
-
rs11816693
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:131099436 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAAGAACGTTAAAAATAAACTGAAG[A/G]TAAGTGTTTACGCAAACATAATAAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TCERG1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10765035] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TCERG1L
- Gene Name
- transcription elongation regulator 1 like
- Gene
- TCERG1L-AS1
- Gene Name
- TCERG1L antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details