Product Details

SNP ID
rs890528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141059921 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACCAGTCTCAGAGAGATTAAGT[C/T]CTGGCCTAGGGTCCAACAGACAGGT
Phenotype
MIM: 611660
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPSB4 PubMed Links

Gene Details

Gene
SPSB4
Gene Name
splA/ryanodine receptor domain and SOCS box containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080862.2 Intron NP_543138.1
XM_017007509.1 Intron XP_016862998.1

View Full Product Details