Product Details

SNP ID
rs1109088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138435953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGCAAACCCTTACTGTCTCTCC[A/G]GTGCTGGGGCTCTTCGTTTTCAGAT
Phenotype
MIM: 616692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESYT3 PubMed Links

Gene Details

Gene
ESYT3
Gene Name
extended synaptotagmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322831.1 Intron NP_001309760.1
NM_001322834.1 Intron NP_001309763.1
NM_031913.4 Intron NP_114119.2
XM_017007307.1 Intron XP_016862796.1

View Full Product Details