Product Details
- SNP ID
-
rs1010187
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139005331 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGCCAGCCAACCAACCAACCCA[C/T]CCAAACAACAGTAATCCAACTGCCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRR23A
PubMed Links
Gene Details
- Gene
- PRR23A
- Gene Name
- proline rich 23A
View Full Product Details