Product Details

SNP ID
rs1533501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:141778308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGGTGGACATGGGGGCCCTGGA[C/T]AACCTGATCGCCAACACCGCCTACC
Phenotype
MIM: 606987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRK7 PubMed Links

Gene Details

Gene
GRK7
Gene Name
G protein-coupled receptor kinase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139209.2 2850 Silent Mutation GAC,GAT D8D NP_631948.1
XM_017005717.1 2850 Silent Mutation GAC,GAT D8D XP_016861206.1

View Full Product Details