Product Details

SNP ID
rs1511802
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:186229652 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTAGGTTTCAGAAAGTTTACTCCA[C/T]AGTTGACTTTTTCTACCAGCTAATA
Phenotype
MIM: 229000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLKB1 PubMed Links

Gene Details

Gene
KLKB1
Gene Name
kallikrein B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000892.4 Intron NP_000883.2
NM_001318394.1 Intron NP_001305323.1
NM_001318396.1 Intron NP_001305325.1
XM_011531930.2 Intron XP_011530232.1
XM_017008181.1 Intron XP_016863670.1
XM_017008182.1 Intron XP_016863671.1
XM_017008183.1 Intron XP_016863672.1
XM_017008184.1 Intron XP_016863673.1

View Full Product Details