Product Details

SNP ID
rs1532485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180602193 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTCTTCCGCTTCCTGTACCACCC[G/T]GCTCAAGTAGCGGACACGGAACAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TTC14 PubMed Links

Gene Details

Gene
TTC14
Gene Name
tetratricopeptide repeat domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042601.2 64 UTR 5 NP_001036066.1
NM_001288582.1 64 UTR 5 NP_001275511.1
NM_133462.3 64 UTR 5 NP_597719.1
XM_011512439.2 64 Intron XP_011510741.1
XM_017005739.1 64 UTR 5 XP_016861228.1
XM_017005740.1 64 UTR 5 XP_016861229.1

View Full Product Details