Product Details

SNP ID
rs1170650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179400089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAGAAACCAGTATTTTGTGCTA[A/G]GGAAGGGAAGAAATACTACAAAATG
Phenotype
MIM: 610863 MIM: 608506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNB4 PubMed Links

Gene Details

Gene
GNB4
Gene Name
G protein subunit beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021629.3 2348 UTR 3 NP_067642.1
XM_005247692.2 2348 UTR 3 XP_005247749.1
XM_006713721.2 2348 UTR 3 XP_006713784.1
Gene
MFN1
Gene Name
mitofusin 1
There are no transcripts associated with this gene.

View Full Product Details