Product Details

SNP ID
rs13634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:16161586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTCCTTTTGTTGGTAAAGGGAGA[C/T]GGAAAAAATAATTCTAAAATGGGGC
Phenotype
MIM: 612758
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAPT1 PubMed Links

Gene Details

Gene
TAPT1
Gene Name
transmembrane anterior posterior transformation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153365.2 4606 UTR 3 NP_699196.2
XM_005248139.2 4606 UTR 3 XP_005248196.1
XM_005248140.3 4606 UTR 3 XP_005248197.1
XM_011513812.2 4606 UTR 3 XP_011512114.1
XM_011513815.2 4606 UTR 3 XP_011512117.1
XM_011513816.2 4606 UTR 3 XP_011512118.1
XM_011513817.2 4606 UTR 3 XP_011512119.1
XM_017007876.1 4606 UTR 3 XP_016863365.1

View Full Product Details