Product Details

SNP ID
rs3182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:3508871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCGAGACTGGGAATAAGATTAGC[A/G]AGCGCCGGCTCACCTCACCAGTGCA
Phenotype
MIM: 610285 MIM: 104225
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOK7 PubMed Links

Gene Details

Gene
DOK7
Gene Name
docking protein 7
There are no transcripts associated with this gene.

Gene
LRPAP1
Gene Name
LDL receptor related protein associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002337.3 5262 UTR 3 NP_002328.1

View Full Product Details