Product Details
- SNP ID
-
rs846238
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:2476794 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTATTTGCAGGTGCAATTATATA[A/G]CACAGTACAGCCTAAAACTTTTTTT
- Phenotype
-
MIM: 602850
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF4
PubMed Links
Gene Details
- Gene
- RNF4
- Gene Name
- ring finger protein 4
View Full Product Details