Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018140.3 | 7419 | Intron | NP_060610.2 | ||
XM_005248322.3 | 7419 | Intron | XP_005248379.1 | ||
XM_011514063.1 | 7419 | Intron | XP_011512365.1 | ||
XM_011514064.2 | 7419 | Intron | XP_011512366.1 | ||
XM_017009626.1 | 7419 | Intron | XP_016865115.1 | ||
XM_017009627.1 | 7419 | Intron | XP_016865116.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_007030.2 | 7419 | UTR 3 | NP_008961.1 | ||
XM_005248237.3 | 7419 | UTR 3 | XP_005248294.2 | ||
XM_017008993.1 | 7419 | UTR 3 | XP_016864482.1 | ||
XM_017008994.1 | 7419 | UTR 3 | XP_016864483.1 |