Product Details

SNP ID
rs1459196
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:64722976 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCACTATCATGAATTCACCTGCAGC[T/C]GTATCTATAGGTTATCAATTTGAAA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FAM159B PubMed Links

Gene Details

Gene
FAM159B
Gene Name
family with sequence similarity 159 member B
There are no transcripts associated with this gene.

Gene
SREK1IP1
Gene Name
SREK1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173829.3 2160 UTR 3 NP_776190.1
XM_011543344.2 2160 UTR 3 XP_011541646.1

View Full Product Details