Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001113561.2 | Intron | NP_001107033.1 | |||
NM_001323291.1 | Intron | NP_001310220.1 | |||
NM_001323292.1 | Intron | NP_001310221.1 | |||
NM_178532.4 | Intron | NP_848627.1 | |||
XM_017009383.1 | Intron | XP_016864872.1 | |||
XM_017009384.1 | Intron | XP_016864873.1 | |||
XM_017009385.1 | Intron | XP_016864874.1 | |||
XM_017009386.1 | Intron | XP_016864875.1 | |||
XM_017009387.1 | Intron | XP_016864876.1 | |||
XM_017009388.1 | Intron | XP_016864877.1 | |||
XM_017009389.1 | Intron | XP_016864878.1 |