Product Details
- SNP ID
-
rs1065358
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44220153 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGTGCCACACGCGGGTCTCCTCA[A/G]ACTGGCTGGTGCGGGGCCGGCCCTG
- Phenotype
-
MIM: 607707
MIM: 606209
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CAMK2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77158048] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAMK2B
- Gene Name
- calcium/calmodulin dependent protein kinase II beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001220.4 |
2125 |
Missense Mutation |
TCT,TTT |
S637F |
NP_001211.3 |
NM_001293170.1 |
2125 |
Missense Mutation |
TCT,TTT |
S513F |
NP_001280099.1 |
NM_172078.2 |
2125 |
Missense Mutation |
TCT,TTT |
S513F |
NP_742075.1 |
NM_172079.2 |
2125 |
Missense Mutation |
TCT,TTT |
S489F |
NP_742076.1 |
NM_172080.2 |
2125 |
Missense Mutation |
TCT,TTT |
S488F |
NP_742077.1 |
NM_172081.2 |
2125 |
Missense Mutation |
TCT,TTT |
S474F |
NP_742078.1 |
NM_172082.2 |
2125 |
Missense Mutation |
TCT,TTT |
S463F |
NP_742079.1 |
NM_172083.2 |
2125 |
Missense Mutation |
TCT,TTT |
S450F |
NP_742080.1 |
NM_172084.2 |
2125 |
Missense Mutation |
TCT,TTT |
S420F |
NP_742081.1 |
XM_005249862.3 |
2125 |
Missense Mutation |
TCT,TTT |
S498F |
XP_005249919.1 |
XM_005249864.3 |
2125 |
Missense Mutation |
TCT,TTT |
S465F |
XP_005249921.1 |
XM_006715781.2 |
2125 |
Missense Mutation |
TCT,TTT |
S597F |
XP_006715844.1 |
XM_006715784.2 |
2125 |
Missense Mutation |
TCT,TTT |
S489F |
XP_006715847.1 |
XM_011515547.1 |
2125 |
Missense Mutation |
TCT,TTT |
S680F |
XP_011513849.1 |
XM_011515549.1 |
2125 |
Missense Mutation |
TCT,TTT |
S665F |
XP_011513851.1 |
XM_011515550.1 |
2125 |
Missense Mutation |
TCT,TTT |
S656F |
XP_011513852.1 |
XM_011515551.1 |
2125 |
Missense Mutation |
TCT,TTT |
S656F |
XP_011513853.1 |
XM_011515552.1 |
2125 |
Missense Mutation |
TCT,TTT |
S655F |
XP_011513854.1 |
XM_011515553.1 |
2125 |
Missense Mutation |
TCT,TTT |
S642F |
XP_011513855.1 |
XM_011515554.1 |
2125 |
Missense Mutation |
TCT,TTT |
S640F |
XP_011513856.1 |
XM_011515555.1 |
2125 |
Missense Mutation |
TCT,TTT |
S637F |
XP_011513857.1 |
XM_011515556.1 |
2125 |
Missense Mutation |
TCT,TTT |
S632F |
XP_011513858.1 |
XM_011515557.1 |
2125 |
Missense Mutation |
TCT,TTT |
S631F |
XP_011513859.1 |
XM_011515558.2 |
2125 |
Intron |
|
|
XP_011513860.1 |
XM_011515559.1 |
2125 |
Missense Mutation |
TCT,TTT |
S551F |
XP_011513861.1 |
XM_017012660.1 |
2125 |
Missense Mutation |
TCT,TTT |
S641F |
XP_016868149.1 |
XM_017012661.1 |
2125 |
Missense Mutation |
TCT,TTT |
S627F |
XP_016868150.1 |
XM_017012662.1 |
2125 |
Missense Mutation |
TCT,TTT |
S574F |
XP_016868151.1 |
XM_017012663.1 |
2125 |
Missense Mutation |
TCT,TTT |
S527F |
XP_016868152.1 |
XM_017012664.1 |
2125 |
Missense Mutation |
TCT,TTT |
S473F |
XP_016868153.1 |
XM_017012665.1 |
2125 |
Missense Mutation |
TCT,TTT |
S464F |
XP_016868154.1 |
XM_017012666.1 |
2125 |
Missense Mutation |
TCT,TTT |
S384F |
XP_016868155.1 |
XM_017012667.1 |
2125 |
Missense Mutation |
TCT,TTT |
S369F |
XP_016868156.1 |
XM_017012668.1 |
2125 |
Missense Mutation |
TCT,TTT |
S360F |
XP_016868157.1 |
XM_017012669.1 |
2125 |
Missense Mutation |
TCT,TTT |
S360F |
XP_016868158.1 |
XM_017012670.1 |
2125 |
Missense Mutation |
TCT,TTT |
S345F |
XP_016868159.1 |
- Gene
- YKT6
- Gene Name
- YKT6 v-SNARE homolog (S. cerevisiae)
There are no transcripts associated with this gene.
View Full Product Details