Product Details
- SNP ID
-
rs1190637
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:104954404 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGAAGGCACAGAAAAAACAGTGC[C/T]ATATTCTTAAATTGGTGAGTGATTA
- Phenotype
-
MIM: 611044
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LIN28B
PubMed Links
Gene Details
- Gene
- LIN28B
- Gene Name
- lin-28 homolog B
View Full Product Details