Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001143940.1 | Intron | NP_001137412.1 | |||
NM_016571.2 | Intron | NP_057655.2 | |||
XM_011535889.2 | Intron | XP_011534191.1 | |||
XM_011535892.2 | Intron | XP_011534194.1 | |||
XM_017010929.1 | Intron | XP_016866418.1 | |||
XM_017010930.1 | Intron | XP_016866419.1 | |||
XM_017010931.1 | Intron | XP_016866420.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003463.4 | Intron | NP_003454.1 | |||
XM_011536111.1 | Intron | XP_011534413.1 | |||
XM_011536112.1 | Intron | XP_011534414.1 | |||
XM_017011270.1 | Intron | XP_016866759.1 | |||
XM_017011271.1 | Intron | XP_016866760.1 | |||
XM_017011272.1 | Intron | XP_016866761.1 |