Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042762.2 | 2851 | UTR 3 | NP_001036227.1 | ||
NM_001287492.1 | 2851 | UTR 3 | NP_001274421.1 | ||
NM_001287493.1 | 2851 | UTR 3 | NP_001274422.1 | ||
NM_001287494.1 | 2851 | UTR 3 | NP_001274423.1 | ||
NM_001287495.1 | 2851 | UTR 3 | NP_001274424.1 | ||
NM_001287496.1 | 2851 | UTR 3 | NP_001274425.1 | ||
NM_022116.4 | 2851 | UTR 3 | NP_071399.2 | ||
XM_011515470.2 | 2851 | UTR 3 | XP_011513772.1 | ||
XM_017012501.1 | 2851 | UTR 3 | XP_016867990.1 | ||
XM_017012502.1 | 2851 | UTR 3 | XP_016867991.1 |