Product Details

SNP ID
rs7842651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23431382 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGGAACAGCACACACAGACACTC[A/G]CACAAAACAAACTCCACCTAAAAAA
Phenotype
MIM: 607577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENTPD4 PubMed Links

Gene Details

Gene
ENTPD4
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128930.2 3643 UTR 3 NP_001122402.1
NM_004901.4 3643 UTR 3 NP_004892.1

View Full Product Details